Learn More
Alkali Scientific Flexible Sealing Mat for Any 96 Well PCR Plates, Reuseable, 10/pk
SDP
Catalog No. 502388920
Encompass_Preferred
This reusable mat provides an airtight seal for all standard 96 well PCR plates, ensuring sample integrity and preventing cross-contamination. Made from high-quality, durable materials, it withstands repeated thermal cycling and autoclaving, making it both cost-effective and environmentally friendly. Its flexible design allows for easy application and removal, while maintaining a secure fit. Enhance your lab's efficiency and reliability with this versatile sealing mat.
Catalog No. 50-238-8920 Supplier Alkali Scientific Supplier No. PC50
This reusable mat provides an airtight seal for all standard 96 well PCR plates, ensuring sample integrity and preventing cross-contamination. Made from high-quality, durable materials, it withstands repeated thermal cycling and autoclaving, making it both cost-effective and environmentally friendly. Its flexible design allows for easy application and removal, while maintaining a secure fit. Enhance your lab's efficiency and reliability with this versatile sealing mat.