Learn More
Chemglass Life Sciences Flexible Drive Shaft, 5/16" End Fittings, One Male, One Female
SDP
Catalog No. CG204701
Encompass_Preferred
Flexible Drive Shaft, 5/16" End Fittings, One Male, One Female. Flexible drive shaft permits the drive motor to be remotely mounted thereby providing for easier access to glassware components. Shaft is 48" in length and can be operated at speeds to 2,000 RPM. Heavy duty 0.250" diameter steel flexible shaft in encased in vinyl with steel end fittings. Can be operated bent at a 12" minimum radius and still have a torque rating of 21 in-lbs. The shaft has 5/16" end fittings, one male and one female. Adapters are available for use with other size motor shafts.
Catalog No. CG204701 Supplier Chemglass Life Sciences Supplier No. CG204701
Flexible Drive Shaft, 5/16" End Fittings, One Male, One Female. Flexible drive shaft permits the drive motor to be remotely mounted thereby providing for easier access to glassware components. Shaft is 48" in length and can be operated at speeds to 2,000 RPM. Heavy duty 0.250" diameter steel flexible shaft in encased in vinyl with steel end fittings. Can be operated bent at a 12" minimum radius and still have a torque rating of 21 in-lbs. The shaft has 5/16" end fittings, one male and one female. Adapters are available for use with other size motor shafts.