Learn More
Ace Glass, Inc. Flexible shaft, 91.4cm (36in), complete unit
SDP
Catalog No. 509558234
Encompass
91.4cm (36) fully flexible drive stirring shaft facilitates isolation of the stirrer motor from the reactor. Reactor end consists of a detachable handpiece with a 1/4 collet and 8mm pin adapter. Maximums: 14,000rpm, 24 in-lbs torque.
Catalog No. 50-955-8234 Supplier Ace Glass, Inc. Supplier No. 808130
91.4cm (36) fully flexible drive stirring shaft facilitates isolation of the stirrer motor from the reactor. Reactor end consists of a detachable handpiece with a 1/4 collet and 8mm pin adapter. Maximums: 14,000rpm, 24 in-lbs torque.