Learn More
Stoelting™ Flexible Shaft Drill, Hang-up Style
SDP
Catalog No. 10001015 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Hang-up style
Catalog No. Model
10-001-015 Hang-up style
1 options
Catalog No. 10-001-015 Supplier Stoelting™ Supplier No. 58650
Add to Cart
Add to Cart
Stoelting™ Flexible Shaft Drill features a powerful motor and flexible shaft that offers very low vibration and noise for long-term, reliable performance.
Motor may be suspended from the optional 58652 Motor Support with Table Clamp.
Specifications
For Use With (Application) Neuroscience
Model Hang-up style
Includes Drill bits