Learn More
Electron Microscopy Sciences Flexible Spatulas, Stainless Steel w/Wooden Handle
SDP
Supplier:  Electron Microscopy Sciences 63321
Encompass_Preferred
This is a sword type spatula, made from stainless steel with a wooden handle. This spatula is very thin and has bendability toward the end and very stiff towards the handle. It offers flexibility when working with soft materials, such as embedding waxes. The overall length is 6 3/4".
Catalog No. 50-322-28