Learn More
MSC PEC Tools Flexible Steel Rule, Stainless Steel, 24 in., 3R Graduation, 1 ea.
Catalog No. 5000508876
Encompass
The 24 in. flexible steel rule is a high-quality precision measurement tool designed for versatility and durability in demanding industrial and metalworking environments. Constructed from fully hardened and tempered stainless steel, this tool features a satin chrome finish that reduces glare for improved readability in various lighting conditions.
  • Flexible body allows for ease of use
  • Blade is fully hardened and tempered
  • Satin chrome finish reduces glare
  • Precisely etched and ink-filled 3R graduations
  • Width of 0.750 in. and thickness of 0.040 in.
  • Made in the USA
Catalog No. 50-005-08876 Supplier MSC Supplier No. 01470632
The 24 in. flexible steel rule is a high-quality precision measurement tool designed for versatility and durability in demanding industrial and metalworking environments. Constructed from fully hardened and tempered stainless steel, this tool features a satin chrome finish that reduces glare for improved readability in various lighting conditions.
  • Flexible body allows for ease of use
  • Blade is fully hardened and tempered
  • Satin chrome finish reduces glare
  • Precisely etched and ink-filled 3R graduations
  • Width of 0.750 in. and thickness of 0.040 in.
  • Made in the USA