Learn More
MSC PEC Tools Flexible Steel Rule, Metric, 450 mm, 1 ea
Catalog No. 5000513433
Encompass
This flexible steel rule is designed for precise measurement and layout tasks in industrial, workshop, or maritime environments. Constructed from spring-tempered stainless steel, the tool features a durable design that offers reliable recovery and resistance to rust and pitting.
  • Features metric graduations of 1 mm and 0.5 mm on both sides
  • Provides a flexible body to facilitate measuring curved surfaces
  • Manufactured from fully hardened and tempered material for enhanced durability
  • Precisely etched and ink-filled markings ensure high readability
  • Conforms to EEC-Class 1 accuracy standards
Catalog No. 50-005-13433 Supplier MSC Supplier No. 01470723
This flexible steel rule is designed for precise measurement and layout tasks in industrial, workshop, or maritime environments. Constructed from spring-tempered stainless steel, the tool features a durable design that offers reliable recovery and resistance to rust and pitting.
  • Features metric graduations of 1 mm and 0.5 mm on both sides
  • Provides a flexible body to facilitate measuring curved surfaces
  • Manufactured from fully hardened and tempered material for enhanced durability
  • Precisely etched and ink-filled markings ensure high readability
  • Conforms to EEC-Class 1 accuracy standards