Learn More
Hillrom FlexiPort™ Reusable Blood Pressure Cuff
Catalog No. 22355156
Change view
Click to view available options
Size:
09 Child
10 Small Adult
11 Adult
12 Large Adult
Includes:
1 Tube, Male Screw Connector
2 Tubes, Tri-Purpose Connector, Inflation Bulb and Valve
No Tubes or Connectors
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes
22-355-156 09 Child No Tubes or Connectors
22-355-157 10 Small Adult No Tubes or Connectors
22-355-158 11 Adult No Tubes or Connectors
22-355-159 11 Adult 1 Tube, Male Screw Connector
22-355-160 11 Adult 2 Tubes, Tri-Purpose Connector, Inflation Bulb and Valve
22-355-161 12 Large Adult 2 Tubes, Tri-Purpose Connector, Inflation Bulb and Valve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-355-156 Supplier Hillrom Supplier No. WEL REUSE09
Add to Cart
Add to Cart
This item is not returnable. View return policy
Invest in an easy-to-use and affordable reusable blood pressure cuff solution. The single-point FlexiPort snap connection was designed to simplify connectivity, reducing the hassle of needing multiple cuff tubes and connectors. Easily size cuffs up or down for each patient.
  • FlexiPort technology enables every cuff to be used as a one- or two-tube cuff
  • Port can be rotated for improved patient comfort
  • Materials do not contain natural rubber latex
  • Meet all latest clinical guidelines for proper fit from AAMI and AHA
  • Primary sizes are color-coded to make it easy to find the proper cuff size
  • Up to 13 sizes, ranging from small infant blood pressure cuffs through large adult blood pressure cuffs (including extra-large, thigh and long cuffs)
Specifications
Size 09 Child
For Use With (Equipment) Blood Pressure Devices and Patient Monitors
Includes No Tubes or Connectors
Latex Free Yes
Product Type Blood Pressure Cuff
Width (Metric) Cuff 15 to 21 cm