Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interscience FlexiPump Pro
SDP

Catalog No. 16949058 Shop All Interscience Products
Change view
Click to view available options
Flow Rate:
3 mL/min. to 2.5 L/min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
16-949-058 3 mL/min. to 2.5 L/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-949-058 Supplier Interscience Supplier No. 562000
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

FlexiPump Pro is a hight-tech peristaltic dispensing pump for culture media preparation with traceability.

FlexiPump Pro is a peristaltic pump with enhanced accuracy for the dispensing of culture media, agar and any type of diluent. Compact and ergonomic, it comes with a user-friendly interface. Traceability is guaranteed by printer and USB connection.

  • Dispensing mode: Continuous - Dose - Multi-Dose
  • Dose volume: From 50 μL to 99.9 L
  • Flow rate: Up to 2.5 L/min.
  • Connection: USB, RS232, Jack 3.5 mm

Specifications

Control Type Analog and Digital
Display Type Digital
Enclosure Rating IP31
Flow Rate 3 mL/min. to 2.5 L/min.
Motor Speed 10 to 550 rpm
Reversible No
Model FlexiPump Pro
Voltage 100/240 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.