Learn More
Med Vet International Flexira Chewable Tablets For Dogs, 227mg, 60 Count
Catalog No. 502827958
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Flexira (firocoxib) Chewable Tablets are indicated for the control of pain and inflammation associated with osteoarthritis and for the control of postoperative pain and inflammation associated with soft-tissue and orthopedic surgery in dogs. BIOEQUIVALENT: FLEXIRA (firocoxib) Chewable Tablets are bioequivalent to Previcox (firocoxib) Chewable Tablets and deliver equivalent absorption and depletion for effective pain relief.* Previcox is a registered trademark of Boehringer Ingelheim Animal Health USA Inc. Pivetal Flexira (firocoxib) Chewable Tablets are not manufactured or distributed by Boehringer Ingelheim Animal Health USA Inc.
Catalog No. 50-282-7958 Supplier Med Vet International Supplier No. RXFLEXIRA22760
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Flexira (firocoxib) Chewable Tablets are indicated for the control of pain and inflammation associated with osteoarthritis and for the control of postoperative pain and inflammation associated with soft-tissue and orthopedic surgery in dogs. BIOEQUIVALENT: FLEXIRA (firocoxib) Chewable Tablets are bioequivalent to Previcox (firocoxib) Chewable Tablets and deliver equivalent absorption and depletion for effective pain relief.* Previcox is a registered trademark of Boehringer Ingelheim Animal Health USA Inc. Pivetal Flexira (firocoxib) Chewable Tablets are not manufactured or distributed by Boehringer Ingelheim Animal Health USA Inc.