Learn More
Cole-Parmer Argos Technologies FlexiRoll Digital Tube Roller Shaker; 220 VAC
Catalog No. 501960337 Shop All Cole-Parmer Products
Encompass_Preferred
Use these compact digital roller shakers with a variety of bottles, tubes, and flasks with minimal aeration. You also can double your capacity and save precious lab space by stacking two units.
Catalog No. 50-196-0337 Supplier Cole-Parmer Supplier No. 0439745
Use these compact digital roller shakers with a variety of bottles, tubes, and flasks with minimal aeration. You also can double your capacity and save precious lab space by stacking two units.