Learn More
REPLIGEN CORPORATION Repligen FlexKit Wye MPX GI, Single-Use Design, Reduced Contamination Risk, MOQ 10 units
SDP
Catalog No. NC2210975
Encompass
The Repligen FlexKit Wye MPX GI is a single-use assembly designed for bioprocessing fluid transfer. With its Wye connector, it ensures efficient and secure fluid branching while minimizing contamination risks. The robust design with overmolded connections optimizes fluid transfer and enhances process integrity, making it ideal for critical applications. This product stands out by eliminating the need for cleaning and sterilization, thereby reducing turnaround times.
This item is not returnable. View return policy
Catalog No. NC2210975 Supplier REPLIGEN CORPORATION Supplier No. FXK00010956
The Repligen FlexKit Wye MPX GI is a single-use assembly designed for bioprocessing fluid transfer. With its Wye connector, it ensures efficient and secure fluid branching while minimizing contamination risks. The robust design with overmolded connections optimizes fluid transfer and enhances process integrity, making it ideal for critical applications. This product stands out by eliminating the need for cleaning and sterilization, thereby reducing turnaround times.