Learn More
Electron Microscopy Sciences Flexwell™ Flexplate, 96 Square Wells
SDP
Catalog No. 5034599
Encompass_Preferred
FlexWell removable incubation chambers for microarrays using ONCYTE slides. Silicone FlexWell forms well on slides using a clean release adhesive to isolate up to 16 specimens per slide. Slides and FlexWell fit into a reusable slide tray organizing wells into the footprint of a SBS compliant microtiter plate. Gasket may be sealed for mixing and to prevent evaporation. Measures 7.25 x 7.25 mm, 9 mm well spacing, 4 mm Depth. Adhesive on Side.
Catalog No. 50-345-99 Supplier Electron Microscopy Sciences Supplier No. 6348555
Add to Cart
Add to Cart
FlexWell removable incubation chambers for microarrays using ONCYTE slides. Silicone FlexWell forms well on slides using a clean release adhesive to isolate up to 16 specimens per slide. Slides and FlexWell fit into a reusable slide tray organizing wells into the footprint of a SBS compliant microtiter plate. Gasket may be sealed for mixing and to prevent evaporation. Measures 7.25 x 7.25 mm, 9 mm well spacing, 4 mm Depth. Adhesive on Side.