Learn More
Molecular Devices Molecular Devices Validation Plate, Luminescence, Microplate, Recertifiable
Catalog No. 5000014362
Encompass_Preferred
The SpectraTest LM1 Luminescence Validation Plate from Molecular Devices provides a standardized reference for verifying luminescence performance of microplate readers. It enables routine performance checks and supports annual recertification to maintain calibration.
  • Enables background noise and spike testing.
  • Measures crosstalk between adjacent wells.
  • Detects left-to-right positional bias across the plate.
  • Supports annual recertification and calibration by an accredited laboratory.
  • Intended for verification of luminescence detection performance in microplate readers.
Catalog No. 50-000-14362 Supplier Molecular Devices Supplier No. 02006186
Add to Cart
Add to Cart
Request a Quote
The SpectraTest LM1 Luminescence Validation Plate from Molecular Devices provides a standardized reference for verifying luminescence performance of microplate readers. It enables routine performance checks and supports annual recertification to maintain calibration.
  • Enables background noise and spike testing.
  • Measures crosstalk between adjacent wells.
  • Detects left-to-right positional bias across the plate.
  • Supports annual recertification and calibration by an accredited laboratory.
  • Intended for verification of luminescence detection performance in microplate readers.