Learn More
Molecular Devices Molecular Devices Pipettor Head, 16-Channel, 384-Well, FlexStation 3 Compatible
Catalog No. 5000014624
Encompass_Preferred
The 16-channel pipettor head is an accessory for the FlexStation 3 multi-mode microplate reader that provides automated, simultaneous aspiration and dispense across 16 channels for high-throughput 384-well assays. It enables rapid plate-to-plate reagent transfer and precise reagent addition for fast kinetic and end-point assays.
  • Provides 16-channel automated liquid handling for rapid reagent transfers.
  • Designed for use with 384-well compound and assay plates.
  • Uses disposable 384-well, 30 μL pipette tips to minimize cross-contamination.
  • Enables timed reagent addition for kinetic assays with millisecond precision.
  • Removable for cleaning and service by trained personnel.
Catalog No. 50-000-14624 Supplier Molecular Devices Supplier No. 02006183
Add to Cart
Add to Cart
Request a Quote
The 16-channel pipettor head is an accessory for the FlexStation 3 multi-mode microplate reader that provides automated, simultaneous aspiration and dispense across 16 channels for high-throughput 384-well assays. It enables rapid plate-to-plate reagent transfer and precise reagent addition for fast kinetic and end-point assays.
  • Provides 16-channel automated liquid handling for rapid reagent transfers.
  • Designed for use with 384-well compound and assay plates.
  • Uses disposable 384-well, 30 μL pipette tips to minimize cross-contamination.
  • Enables timed reagent addition for kinetic assays with millisecond precision.
  • Removable for cleaning and service by trained personnel.