Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont Industrial FlexTech ANSI A4 Cut Resistant Gloves

Catalog No. 17010052 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-052 Medium
17-010-051 Small
17-010-053 Large
17-010-054 X-Large
17-010-055 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-010-052 Supplier Wells Lamont Industrial Supplier No. Y9258M
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

FlexTech ANSI A4 Cut Resistant Gloves with a foam nitrile palm and ESD properties to protect equipment and processes.

  • Anti-static properties that rapidly neutralize the electric discharge for the worker and ESD properties that protect product equipment and workpieces
  • The ultra-lightweight liner provides excellent dexterity as an 18 gauge thin glove with an ANSI Cut Level A4 protection to reduce hand injuries and increased abrasion resistance and durability for prolonged use
  • Foam nitrile palm for grip and touchscreen compatibility
  • Offers ANSI 4 puncture and abrasion resistance

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance EN 16350:2014, EN 1149-2:1997
Coated Area Palm
Applications Industrial
Coating Material Nitrile Foam
Color Gray
Cut Resistance Rating ANSI A4
Disposable Reusable
Exterior Finish Fully Textured
Material Nylon, HPPE, Fiberglass, Lycra, Stainless Steel
Product Type Cut Resistant Glove
Size Medium
For Use With (Application) Metal Stamping, Electronics Assembly, Small Parts Handling and Assembly
Model Y9258
Resistant To Cut Resistant
Yarn Gauge 18
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.