Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Cut Resistant Gloves with Nitrile Coating

Catalog No. 19086710 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-086-710 2X-Large
19-086-706 Small
19-086-707 Medium
19-086-708 Large
19-086-709 X-Large
19-086-705 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-086-710 Supplier Wells Lamont™ Supplier No. Y9216XXL
  • $556.50
    $253.65 / Dozen of 12

    Save $302.85

    54% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $5,810.00
    $2,648.65 / Case of 12 DZ

    Save $3,161.35

    54% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FlexTech™ Cut Resistant Gloves with Nitrile Coating offer ANSI A7 cut resistance, dexterity, and an excellent grip, especially in wet and oily conditions.

  • Seamless machine knit cut resistant shell provides all-day comfort and protection
  • Sandy nitrile palm coating delivers improved grip in wet and oily applications
  • Machine washable for longer service life
  • 13 Gauge
  • ANSI Abrasion Level 5
  • ANSI Puncture Level 5
  • ANSI Cut Level A7
  • Sold by the dozen pair

Specifications

Construction Seamless Knit
Coating Material Nitrile
Color Black, Gray
Cut Resistance Rating ANSI A7
Hand Specific
Product Type Cut-resistant Gloves
Size 2X-Large
For Use With (Application) For improved grip in wet and oily applications.
Glove Cuff Style Gauntlet
Model Y9216
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.