Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Polyurethane Coated Nylon Work Gloves

Catalog No. 19148723 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-723 X-Large
19-148-719 X-Small
19-148-720 Small
19-148-721 Medium
19-148-722 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-148-723 Supplier Wells Lamont™ Supplier No. Y9277XL
  • $556.24
    $278.65 / Case of 144

    Save $277.59

    50% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FlexTech™ Polyurethane Coated Nylon Work Gloves feature polyurethane coating on the palm and fingers that provides additional protection in high wear, high risk areas of the hand.

  • Nylon gloves with polyurethane palm coating to enhance grip and tactility
  • Charcoal gray fibers hide dirt and grime for extended glove life
  • Seamless design increases dexterity and flexibility
  • Ideal for electronic assembly and small parts handling

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ANSI/ISEA 105-2024 Cut Level A1, ANSI/ISEA 105-2024 Abrasion Level 4, ANSI/ISEA 105-2024 Puncture Level 2, EN 388:2016+A1:2018 4131
Certifications/Compliance (Europe) EN 388:2016+A1:2018 4131
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A1
Exterior Finish Fully Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Large
For Use With (Application) Electronic Assembly, and Small Parts Handling
Gender Unisex
Glove Cuff Style Knit
Grip Design Textured
Series Y9277
Yarn Gauge 13
Product Line FlexTech
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

WARNING: Cancer – www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.