Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Flextech™ Polyurethane Coated Synthetic Knit Gloves

Catalog No. 17200404 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
X-Small
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-404 2X-Large
17-200-400 Small
17-200-401 Medium
17-200-403 X-Large
17-200-399 X-Small
17-200-402 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-200-404 Supplier Wells Lamont™ Supplier No. Y9297W11
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flextech™ Polyurethane Coated Synthetic Knit Gloves are general purpose gloves offering a high level of dexterity, with a polyurethane coating on the palm to provide a non-slip gripping surface.

  • 13 gauge, synthetic knit shell increases dexterity and flexibility
  • Polyurethane coating on the palm enhances grip capabilities
  • Open, uncoated back circulates air to allow hands to breathe
  • Non-slip surface allows workers to grip objects with confidence
  • Silicone-free

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ANSI/ISEA 105-2024 Cut Level A1, ANSI/ISEA 105-2024 Abrasion Level 4, EN 388:2016+A1:2018 3131, Silicone Free
Certifications/Compliance (Europe) EN 388:2016+A1:2018 3131
Construction Seamless Knit
Coating Material Polyurethane
Color White
Exterior Finish Smooth
Hand Specific
Latex Free No
Material Nylon
Product Type Seamless Knit Gloves
Size 2X-Large
Sterility Non-sterile
For Use With (Application) General Purpose Work
Gender Unisex
Glove Cuff Color Yellow
Glove Cuff Style Knit
Series Y9297W
Yarn Gauge 13
Product Line FlexTech
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.