Learn More
Wells Lamont™ Flextech™ Sandy Nitrile-Coated Hi-Vis Heat Resistant Gloves
Catalog No. 17200408 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
2X-Large
Catalog No. Size
17-200-408 2X-Large
1 options
This item is not returnable. View return policy
Catalog No. 17-200-408 Supplier Wells Lamont™ Supplier No. Y9241TXXL
This item is not returnable. View return policy
Gloves are rated ANSI Level 2 for conductive heat to offer protection from temperatures up to 350°F. Sandy nitrile coating on the palm enhances grip in wet or oily conditions.
  • Sandy nitrile coated palm enhances grip capabilities in wet and oily conditions
  • Protection from temperatures up to 350°F (175°C)
  • ANSI Level 2 Conductive Heat per ASTM F1060-08
  • Hi-vis orange shell increases visibility during low-light applications
  • ANSI Cut Level A2 provides cut protection against sharp objects
  • Rated ANSI Level 2 for Conductive Heat
Specifications
Coating Material Sandy Nitrile
Color Hi-vis Orange
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free No
Size 2X-Large
Temperature (English) Max. 350°F
Temperature (Metric) Max 175°C
Glove Cuff Style Knit
Series Y9241T