Learn More
Research Products International Corp Wash Bottle with Flexible Tip, 250ml, 12 per Package
SDP
Catalog No. 50212576
Encompass
Low density polyethylene bottles are translucent for easy viewing of contents. Slim flexible spout delivers a steady stream of liquid by gently squeezing the bottle. Supplied with tip cover for storage. Bottles can be labelled or marked with a felt tip pen to identify contents.
Catalog No. 50-212-576 Supplier Research Products International Corp Supplier No. 248642
Low density polyethylene bottles are translucent for easy viewing of contents. Slim flexible spout delivers a steady stream of liquid by gently squeezing the bottle. Supplied with tip cover for storage. Bottles can be labelled or marked with a felt tip pen to identify contents.