Learn More
Research Products International Corp Wash Bottle with Flexible Tip, 500ml, 6 per Package
SDP
Catalog No. 50212577
Encompass
Low density polyethylene bottles are translucent for easy viewing of contents. Slim flexible spout delivers a steady stream of liquid by gently squeezing the bottle. Supplied with tip cover for storage. Bottles can be labelled or marked with a felt tip pen to identify contents.
Catalog No. 50-212-577 Supplier Research Products International Corp Supplier No. 248643
Add to Cart
Add to Cart
Low density polyethylene bottles are translucent for easy viewing of contents. Slim flexible spout delivers a steady stream of liquid by gently squeezing the bottle. Supplied with tip cover for storage. Bottles can be labelled or marked with a felt tip pen to identify contents.