Learn More
Electron Microscopy Sciences Flexwell™ 16 wells 6.5x6.5 mmx3.2 mmD, Clear silicone
SDP
Catalog No. 5034597
Encompass_Preferred
FlexWell removable incubation chambers for microarrays using ONCYTE slides. Silicone FlexWell forms well on slides using a clean release adhesive to isolate up to 16 specimens per slide. Slides and FlexWell fit into a reusable slide tray organizing wells into the footprint of a SBS compliant microtiter plate. Gasket may be sealed for mixing and to prevent evaporation. 6.5 x 6.5 mm Wells 3.2mm Depth 25 x 75 mm OD.
Catalog No. 50-345-97 Supplier Electron Microscopy Sciences Supplier No. 6348551
FlexWell removable incubation chambers for microarrays using ONCYTE slides. Silicone FlexWell forms well on slides using a clean release adhesive to isolate up to 16 specimens per slide. Slides and FlexWell fit into a reusable slide tray organizing wells into the footprint of a SBS compliant microtiter plate. Gasket may be sealed for mixing and to prevent evaporation. 6.5 x 6.5 mm Wells 3.2mm Depth 25 x 75 mm OD.