Learn More
Electron Microscopy Sciences Flexwell™ 64 wells 3.5x3.5 mmx1.8 mmD, Clear Silicone
SDP
Catalog No. 5034594
Encompass_Preferred
FlexWell removable incubation chambers for microarrays using ONCYTE slides. Silicone FlexWell forms well on slides using a clean release adhesive to isolate up to 16 specimens per slide. Slides and FlexWell fit into a reusable slide tray organizing wells into the footprint of a SBS compliant microtiter plate. Gasket may be sealed for mixing and to prevent evaporation. 3.5 x 3.5 mm Wells 1.8mm Depth 25 x 75 mm OD.
Catalog No. 50-345-94 Supplier Electron Microscopy Sciences Supplier No. 6348552
FlexWell removable incubation chambers for microarrays using ONCYTE slides. Silicone FlexWell forms well on slides using a clean release adhesive to isolate up to 16 specimens per slide. Slides and FlexWell fit into a reusable slide tray organizing wells into the footprint of a SBS compliant microtiter plate. Gasket may be sealed for mixing and to prevent evaporation. 3.5 x 3.5 mm Wells 1.8mm Depth 25 x 75 mm OD.