Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2312MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: SPM181. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: U-251-MG cells or Skin. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length Filaggrin protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 26-45kDa (Processed) ;350kDa (Profilaggrin).Chromosome: 1q21.3. Gene Symbol: FLG. Gene ID: 2312. Ensemble: ENSG00000143631. Uniprot: NP_002007.
Catalog No. 50-188-4683