Learn More
Apexbio Technology LLC FLI-06 313967-18-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089864
Encompass_Preferred
FLI-06 (CAS 313967-18-9) is a small molecule inhibitor of the Notch signaling pathway exhibiting an EC50 of 2 3 M Notch signaling regulates cell fate decisions and its dysregulation is implicated in oncogenesis and developmental disorders FLI-06 impedes Notch E-eGFP accumulation and reduces NICD-eGFP production in HeLa cells with effects that are fully reversible upon washout indicating low cytotoxicity Additionally FLI-06 significantly decreases A secretion upstream of - and -secretase cleavage events and disrupts Golgi structure by interfering with early secretory pathway membrane trafficking This compound is valuable for studying Notch signaling secretory pathway dynamics and related cellular processes
Catalog No. 50-000-89864 Supplier Apexbio Technology LLC Supplier No. B30835.1
FLI-06 (CAS 313967-18-9) is a small molecule inhibitor of the Notch signaling pathway exhibiting an EC50 of 2 3 M Notch signaling regulates cell fate decisions and its dysregulation is implicated in oncogenesis and developmental disorders FLI-06 impedes Notch E-eGFP accumulation and reduces NICD-eGFP production in HeLa cells with effects that are fully reversible upon washout indicating low cytotoxicity Additionally FLI-06 significantly decreases A secretion upstream of - and -secretase cleavage events and disrupts Golgi structure by interfering with early secretory pathway membrane trafficking This compound is valuable for studying Notch signaling secretory pathway dynamics and related cellular processes