Learn More
Aviva Systems Biology Corp FLI1 Antibody - middle region (P100882_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100882_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FLI1. Alternate names: EWSR2, SIC-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62761