Learn More
Biomatik Corporation Fli1 Recombinant Antibody
Supplier:  Biomatik Corporation RAC07068
Encompass_Preferred
Fli1 Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to FLI1. The Fli1 Recombinant Antibody has been validated for the following applications: ELISA, WB. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: A Synthesized Peptide Derived From Human Fli1. Applications: ELISA, WB. Target Synonyms: FLI1. Accession Number: Q01543. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6390