Learn More
Medchemexpress LLC Flibanserin-d4-1 | 2122830-91-3 | 99.3% | 394.43 g/mol | C20H17D4F3N4O | 10 MG
SDP
Catalog No. 5000297918
Encompass_Preferred
Flibanserin-d4-1 is a deuterium-labeled analog of flibanserin supplied as a solid research standard for analytical and pharmacokinetic applications. It functions as a 5-HT1A full agonist and a 5-HT2A antagonist and is used as an internal standard or tracer to support quantitative LC-MS and GC-MS methods and metabolic studies.
  • Deuterium-labeled internal standard for analytical assays.
  • High purity (~99.3%).
  • Solid, white to off-white powder form.
  • Molecular weight 394.43 g/mol.
  • Available as a 10 mg pack.
  • Suitable for LC-MS, GC-MS, and pharmacokinetic studies.
Catalog No. 50-002-97918 Supplier Medchemexpress LLC Supplier No. HYA0095S110MG
Flibanserin-d4-1 is a deuterium-labeled analog of flibanserin supplied as a solid research standard for analytical and pharmacokinetic applications. It functions as a 5-HT1A full agonist and a 5-HT2A antagonist and is used as an internal standard or tracer to support quantitative LC-MS and GC-MS methods and metabolic studies.
  • Deuterium-labeled internal standard for analytical assays.
  • High purity (~99.3%).
  • Solid, white to off-white powder form.
  • Molecular weight 394.43 g/mol.
  • Available as a 10 mg pack.
  • Suitable for LC-MS, GC-MS, and pharmacokinetic studies.