Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flibanserin hydrochloride (propan-2-ol) hydrate | 167933-07-5 | MFCD00918402 | 99.0% | 10 MG
SDP

Catalog No. 5000298517
Encompass_Preferred

Flibanserin hydrochloride (propan-2-ol) hydrate is the hydrochloride salt hydrate of flibanserin (BIMT-17), an orally active serotonin 5-HT1A receptor agonist and 5-HT2A receptor antagonist used in laboratory research on mood, anxiety, and hypoactive sexual desire disorder (HSDD). It is supplied as a solid, high-purity material intended for preclinical and in vitro studies; not for human use.

  • Molecular formula: C20H22ClF3N4O·0.5C3H8O·0.5H2O
  • Molecular weight: 465.20 g/mol
  • Purity: 99.04%
  • Physical state: Solid
  • Salt form: Hydrochloride (propan-2-ol) hydrate
  • Typical applications: preclinical pharmacology and receptor binding studies
  • Available pack sizes: research quantities including 10 MG

Catalog No. 50-002-98517 Supplier Medchemexpress LLC Supplier No. HYA0095A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flibanserin hydrochloride (propan-2-ol) hydrate is the hydrochloride salt hydrate of flibanserin (BIMT-17), an orally active serotonin 5-HT1A receptor agonist and 5-HT2A receptor antagonist used in laboratory research on mood, anxiety, and hypoactive sexual desire disorder (HSDD). It is supplied as a solid, high-purity material intended for preclinical and in vitro studies; not for human use.

  • Molecular formula: C20H22ClF3N4O·0.5C3H8O·0.5H2O
  • Molecular weight: 465.20 g/mol
  • Purity: 99.04%
  • Physical state: Solid
  • Salt form: Hydrochloride (propan-2-ol) hydrate
  • Typical applications: preclinical pharmacology and receptor binding studies
  • Available pack sizes: research quantities including 10 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.