Learn More
Medchemexpress LLC Flibanserin (hydrochloride) (propan-2-ol) (hydrate) | 99.0% | C20H22ClF3N4O.0.5C3H8O.0.5H2O | 25 MG
SDP
Catalog No. 5000357861
Encompass_Preferred
Flibanserin hydrochloride (propan-2-ol) hydrate is an orally active serotonin 5-HT1A receptor agonist and 5-HT2A receptor antagonist. It also binds to dopamine D4 receptors, demonstrating anti-depression and anti-anxiety effects. This compound is utilized in research for hypoactive sexual desire disorder (HSDD).
  • Orally active serotonin 5-HT1A receptor agonist
  • 5-HT2A receptor antagonist
  • Binds to dopamine D4 receptors
  • Shows anti-depression and anti-anxiety effects
  • Used in research for hypoactive sexual desire disorder (HSDD)
  • Non-toxic up to 100 μM in cell viability assays
Catalog No. 50-003-57861 Supplier Medchemexpress LLC Supplier No. 25MGHYA0095A
Flibanserin hydrochloride (propan-2-ol) hydrate is an orally active serotonin 5-HT1A receptor agonist and 5-HT2A receptor antagonist. It also binds to dopamine D4 receptors, demonstrating anti-depression and anti-anxiety effects. This compound is utilized in research for hypoactive sexual desire disorder (HSDD).
  • Orally active serotonin 5-HT1A receptor agonist
  • 5-HT2A receptor antagonist
  • Binds to dopamine D4 receptors
  • Shows anti-depression and anti-anxiety effects
  • Used in research for hypoactive sexual desire disorder (HSDD)
  • Non-toxic up to 100 μM in cell viability assays