Learn More
Medchemexpress LLC Flibanserin hydrochloride (propan-2-ol) hydrate | 99.0% | C20H22ClF3N4O.0.5C3H8O.0.5H2O | 5 MG
SDP
Catalog No. 5000432785
Encompass_Preferred
Flibanserin hydrochloride (propan-2-ol) hydrate is an orally active serotonin 5-HT1A receptor agonist and 5-HT2A receptor antagonist. It demonstrates anti-depression and anti-anxiety effects and is used in research for hypoactive sexual desire disorder (HSDD).
  • Orally active serotonin 5-HT1A receptor agonist
  • Serotonin 5-HT2A receptor antagonist
  • Binds to dopamine D4 receptors
  • Exhibits anti-depression and anti-anxiety effects
  • Used in research for hypoactive sexual desire disorder (HSDD)
  • Shows no toxicity potential after oxidative degradation in vitro
  • Activates brain regions involved in sexual motivation
  • Produces anxiolytic effects without locomotor side effects in rat models
Catalog No. 50-004-32785 Supplier Medchemexpress LLC Supplier No. HYA0095A5MG
Flibanserin hydrochloride (propan-2-ol) hydrate is an orally active serotonin 5-HT1A receptor agonist and 5-HT2A receptor antagonist. It demonstrates anti-depression and anti-anxiety effects and is used in research for hypoactive sexual desire disorder (HSDD).
  • Orally active serotonin 5-HT1A receptor agonist
  • Serotonin 5-HT2A receptor antagonist
  • Binds to dopamine D4 receptors
  • Exhibits anti-depression and anti-anxiety effects
  • Used in research for hypoactive sexual desire disorder (HSDD)
  • Shows no toxicity potential after oxidative degradation in vitro
  • Activates brain regions involved in sexual motivation
  • Produces anxiolytic effects without locomotor side effects in rat models