Learn More
Cayman Chemical Flibanserin-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 2748630-46-6, 1 mg
SDP
Catalog No. 502932669
Encompass_Preferred
An internal standard for the quantification of flibanserin by GC- or LC-MS
Catalog No. 50-293-2669 Supplier Cayman Chemical Supplier No. 267791 MG
An internal standard for the quantification of flibanserin by GC- or LC-MS