Learn More
Cayman Chemical Flibanserin-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 2748630-46-6, 5 mg
SDP
Catalog No. 502934034
Encompass_Preferred
An internal standard for the quantification of flibanserin by GC- or LC-MS
Catalog No. 50-293-4034 Supplier Cayman Chemical Supplier No. 267795 MG
An internal standard for the quantification of flibanserin by GC- or LC-MS