Learn More
V&P Scientific Inc MAGNETIC PLATE, Flick 'N Blot Design, 96 Well, Standard or Deep Well, Round or Conical Bottom, 24 Round Posts
Catalog No. NC1427442
Encompass
MAGNETIC BEAD SEPARATION PLATE, Flick 'N Blot Design, 96 Well, Standard or Deep Well Plate, Round or Conical Bottom, 24 Round Posts, 52MGO, NdFeB Magnets, Red Anodized Magnet Frame, 1 Sided Pellet Location, 6.4mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Non-SLAS Footprint, Includes Polypropylene Base with Adjustable Clips to hold Plates and Black O-Ring Grip
Catalog No. NC1427442 Supplier V&P Scientific Inc Supplier No. VP 771HHLA
MAGNETIC BEAD SEPARATION PLATE, Flick 'N Blot Design, 96 Well, Standard or Deep Well Plate, Round or Conical Bottom, 24 Round Posts, 52MGO, NdFeB Magnets, Red Anodized Magnet Frame, 1 Sided Pellet Location, 6.4mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Non-SLAS Footprint, Includes Polypropylene Base with Adjustable Clips to hold Plates and Black O-Ring Grip