Learn More
AdipoGen FliD mAb rec SH1579-B7
SDP
Supplier:  AdipoGen AG27B6322C050
Encompass_Preferred
Recombinant Antibody. Recognizes FliD from Pseudomonas aeruginosa. Applications ELISA, WB. Clone SH1579-B7. Isotype Human IgG1. Formulation Liquid. In PBS. Pathogenic bacteria cause a multitude of deadly human diseases. Pseudomonas aeruginosa is a predominant organism within the hospital environment, an increasingly multidrug-resistant microbe, and the most common gram-negative pathogen causing nosocomial pneumonia in the United States. Nearly all P. aeruginosa infections are associated with compromised host defenses, which may include patients with severe burns, diabetes, cancer, organ transplants or additional immunodeficiencies. Many of these pathogenic bacteria possess flagella, molecular machines responsible for cell motility, adherence to host cells, and pathogenicity.
Catalog No. 50-256-3121