Learn More
Medchemexpress LLC HY-108584 5mg Medchemexpress, Flindokalner CAS:187523-35-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1085845MG
Encompass_Preferred
Medchemexpress, HY-108584 5mg Flindokalner CAS:187523-35-9 Flindokalner (BMS-204352) is a potassium channel modulator. Flindokalner is a positive modulator of all neuronal Kv7 channel subtypes expressed in HEK293 cells. Flindokalner is also a large conductance calcium-activated K channel (BKca) positive modulator. Flindokalner shows a negative modulatory activity at Kv7.1 channels (Ki=3.7 μM), and acts as a negative modulator of GABAA receptors. Flindokalner shows anxiolytic efficacy in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8579