Learn More
Formulatrix FLIO8 20 UL TIP 10 X 96 PCS
SDP
Catalog No. NC2460086
Encompass
Package of FLO i8 F20 Next Level Tips. Contains 10 x 96 pcs (960 pcs) F20 conductive pipette tips with filter. Tip working volume from 0.5 to 20 µL.
Catalog No. NC2460086 Supplier Formulatrix Supplier No. 800001B
Package of FLO i8 F20 Next Level Tips. Contains 10 x 96 pcs (960 pcs) F20 conductive pipette tips with filter. Tip working volume from 0.5 to 20 µL.