Learn More
AdipoGen anti-FLIP (human), mAb (NF6) (100 ug)
SDP
Supplier:  AdipoGen AG20B0056C100
Encompass_Preferred
anti-FLIP (human), mAb (NF6) (100 ug), Adipogen Life Sciences. Monoclonal Antibody. Recognizes short (FLIPS) and long (FLIPL) splice variants of human FLIP. This antibody recognizes an epitope in the N-terminal DED region (aa1-194). Isotype: Mouse IgG1. Clone: NF6. Applications: ICC, IHC, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. FLIP is an apoptosis regulator protein which functions as a crucial link between cell survival and cell death pathways in mammalian cells and acts as an inhibitor of TNFRSF6 mediated apoptosis. A proteolytic fragment (p43) is likely retained in the death-inducing signaling complex (DISC) thereby blocking further recruitment and processing of caspase-8 at the complex. Full length and shorter isoforms have been shown either to induce apoptosis or to reduce TNFRSF-triggered apoptosis. FLIP lacks enzymatic (caspase) activity. FLIP is highly expressed in skeletal muscle, pancreas, heart, kidney, placenta and peripheral blood leukocytes.
Catalog No. 50-168-7227