Learn More
Enzo Life Sciences FLIP (human) monoclonal antibody (7F10) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX8049610100
Encompass_Preferred
Clone: 7F10. Isotype: IgG1. Immunogen: Recombinant human FLIP. Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, Flow Cytometry, IHC, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-0027