Learn More
AdipoGen anti-FLIP, mAb (Dave-2),Monoclonal Antibody
SDP
Supplier:  AdipoGen AG20B0005C100
Encompass_Preferred
anti-FLIP, mAb (Dave-2), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes an epitope (aa 1-200) present in both short (FLIPS) and long (FLIPL) splice variants of human and mouse FLIP. Isotype: Rat IgG2a. Clone: Dave-2. Applications: IP, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide.
Catalog No. 50-114-5671