Learn More
Electron Microscopy Sciences Lyophilization Flipcap Center Tear Seals, 13 mm I.D., 20 mm O.D.
SDP
Catalog No. 501898950
Encompass_Preferred
Aluminum lyophilization seals are unlined. Easy-to-use, no sharp metal edges. Removal can be accomplished with one hand, fit mouth ID 13mm x OD 20mm.
Catalog No. 50-189-8950 Supplier Electron Microscopy Sciences Supplier No. 6032113
Aluminum lyophilization seals are unlined. Easy-to-use, no sharp metal edges. Removal can be accomplished with one hand, fit mouth ID 13mm x OD 20mm.