Learn More
Electron Microscopy Sciences Lyophilization Flipcap Center Tear Seals, 7 mm I.D., 13 mm O.D.
SDP
Catalog No. 501898949
Encompass_Preferred
Aluminum lyophilization seals are unlined. Easy-to-use, no sharp metal edges. Removal can be accomplished with one hand, fit mouth ID 7mm x OD 13mm.
Catalog No. 50-189-8949 Supplier Electron Microscopy Sciences Supplier No. 6032107
Aluminum lyophilization seals are unlined. Easy-to-use, no sharp metal edges. Removal can be accomplished with one hand, fit mouth ID 7mm x OD 13mm.