Learn More
Cole-Parmer FLIPMATE 50 SC0604 .45UM 100PK
Supplier:  Cole-Parmer 3520218
Encompass
Environmental Express FlipMate 50 SC0604 Filtration Assemblies, Certified, PES/PTFE, 0.45um, 50 mL; 100/Pk
Catalog No. NC2309000