Learn More
Research Products International Corp Flipper Junior Work Racks with Lids, Assorted Fluorescent Colors, 5 per Package
SDP
Catalog No. 50212351
Encompass
Flipper Junior Rack is an ideal small working rack designed for sequencing procedures. One side holds 32 x 0.5 ml tubes with additional row of 8 wells for 1.5, 2.0 ml tubes. The reverse side has opposite configuration that holds 32 x 1.5, 2.0 ml tubes and row of 8 wells for 0.5 ml tubes.
Racks are polypropylene and feature alphanumeric indexing for easy identification.
Catalog No. 50-212-351 Supplier Research Products International Corp Supplier No. 145921
Flipper Junior Rack is an ideal small working rack designed for sequencing procedures. One side holds 32 x 0.5 ml tubes with additional row of 8 wells for 1.5, 2.0 ml tubes. The reverse side has opposite configuration that holds 32 x 1.5, 2.0 ml tubes and row of 8 wells for 0.5 ml tubes.
Racks are polypropylene and feature alphanumeric indexing for easy identification.