Learn More
Research Products International Corp Flipper 4-Way Rack for 5 ml Tubes, Assorted Colors, 4 per Case
SDP
Catalog No. 501367531
Encompass
Flipper 4-Way Tube Rack is compatible with 5 ml Tubes. Designed to hold a variety of tube sizes and styles. Racks can be rotated to any of four positions to accommodate:
  • 32 - 12 x 75 or 1.5 ml tubes
  • 12 - 15 ml tubes
  • 32 - 0.5 ml micro-tubes
  • 18 - 5 ml tubes
  • 4 - 50ml tubes
Autoclavable racks are constructed of durable, polypropylene and can be used in water baths and freezers.
Catalog No. 50-136-7531 Supplier Research Products International Corp Supplier No. 167005
Flipper 4-Way Tube Rack is compatible with 5 ml Tubes. Designed to hold a variety of tube sizes and styles. Racks can be rotated to any of four positions to accommodate:
  • 32 - 12 x 75 or 1.5 ml tubes
  • 12 - 15 ml tubes
  • 32 - 0.5 ml micro-tubes
  • 18 - 5 ml tubes
  • 4 - 50ml tubes
Autoclavable racks are constructed of durable, polypropylene and can be used in water baths and freezers.