Learn More
Cytoskeleton, Inc. Flipper-TR Membrane Sensor, Real-Time Measurement, Non-Invasive Fluorescence, 50 nmol
Catalog No. NC2000715
Encompass
The Flipper-TR Membrane Sensor is an advanced fluorogenic probe designed for real-time measurement of plasma membrane tension in live cells. It utilizes Fluorescence Lifetime Imaging Microscopy (FLIM) to provide insight into membrane mechanics, distinguishing it from other probes. Its unique sensing mechanism allows fluorescence only when incorporated into lipid membranes, making it ideal for diverse organisms, including bacteria and plants. This probe enhances mechanobiology studies by simplifying complex methodologies.
Catalog No. NC2000715 Supplier Cytoskeleton, Inc. Supplier No. CYSC020
The Flipper-TR Membrane Sensor is an advanced fluorogenic probe designed for real-time measurement of plasma membrane tension in live cells. It utilizes Fluorescence Lifetime Imaging Microscopy (FLIM) to provide insight into membrane mechanics, distinguishing it from other probes. Its unique sensing mechanism allows fluorescence only when incorporated into lipid membranes, making it ideal for diverse organisms, including bacteria and plants. This probe enhances mechanobiology studies by simplifying complex methodologies.