Learn More
Research Products International Corp Clear Polypropylene Lid for Flipper Micro-Tube Racks, 10 per Case
SDP
Catalog No. 50212350
Encompass
Clear polypropylene lid is perfect for freezer storage. Lid offers ample height to accommodate screw cap micro-tubes. Lids are autoclavable.
Catalog No. 50-212-350 Supplier Research Products International Corp Supplier No. 145920
Clear polypropylene lid is perfect for freezer storage. Lid offers ample height to accommodate screw cap micro-tubes. Lids are autoclavable.