Learn More
Molecular Devices Molecular Devices Assay Kit, Fluorescent Calcium, Bulk, 96, 384, 1536-Well
Catalog No. 5000014404
Encompass_Preferred
The FLIPR Calcium 5 Assay Kit (Bulk) is a homogeneous, dye-based fluorescent assay designed for high-throughput measurement of intracellular calcium flux using FLIPR systems. The bulk configuration supplies reagents for large screening campaigns and supports 96-, 384-, and 1536-well plate formats.
  • Homogeneous dye-based calcium flux assay for adherent and suspension cells.
  • Bulk kit coverage: 100 plates for 96- or 384-well, 150 plates for 1536-well formats.
  • Includes multiple vials of loading reagent; Component B (assay buffer) is not included in bulk kit.
  • Amenable to incubation at 37°C or room temperature, enabling flexible workflows.
  • Probenecid may be required for some cell lines; do not freeze loading buffer with probenecid.
  • Store components at -20°C and 4°C; reagents are stable for six months in original packaging.
Catalog No. 50-000-14404 Supplier Molecular Devices Supplier No. R8186
Add to Cart
Add to Cart
Request a Quote
The FLIPR Calcium 5 Assay Kit (Bulk) is a homogeneous, dye-based fluorescent assay designed for high-throughput measurement of intracellular calcium flux using FLIPR systems. The bulk configuration supplies reagents for large screening campaigns and supports 96-, 384-, and 1536-well plate formats.
  • Homogeneous dye-based calcium flux assay for adherent and suspension cells.
  • Bulk kit coverage: 100 plates for 96- or 384-well, 150 plates for 1536-well formats.
  • Includes multiple vials of loading reagent; Component B (assay buffer) is not included in bulk kit.
  • Amenable to incubation at 37°C or room temperature, enabling flexible workflows.
  • Probenecid may be required for some cell lines; do not freeze loading buffer with probenecid.
  • Store components at -20°C and 4°C; reagents are stable for six months in original packaging.