Learn More
Molecular Devices Molecular Devices FLIPR Calcium 6-QF Assay Kit, Quench-free, 10 plates
Catalog No. 5000014345
Encompass_Preferred
The FLIPR Calcium 6-QF Assay Kit (Explorer kit) is a quench-free, mix-and-read fluorescent assay for measuring intracellular calcium flux in high-throughput plate formats. The Calcium 6-QF formulation provides an increased signal window and is suitable for quench-sensitive targets and multiplexed assays.
  • Quench-free formulation for quench-sensitive targets and multiplexing.
  • Designed for mix-and-read workflows with pre-optimized FLIPR and FlexStation protocols.
  • Sufficient reagents for 10 plates in Explorer kit format.
  • Each reagent vial for Sample and Explorer kits is sufficient for one 96-, 384-, or 1536-well plate.
  • Suitable for endogenous, primary, and stem cell targets with improved signal window and assay robustness.
Catalog No. 50-000-14345 Supplier Molecular Devices Supplier No. R8192
The FLIPR Calcium 6-QF Assay Kit (Explorer kit) is a quench-free, mix-and-read fluorescent assay for measuring intracellular calcium flux in high-throughput plate formats. The Calcium 6-QF formulation provides an increased signal window and is suitable for quench-sensitive targets and multiplexed assays.
  • Quench-free formulation for quench-sensitive targets and multiplexing.
  • Designed for mix-and-read workflows with pre-optimized FLIPR and FlexStation protocols.
  • Sufficient reagents for 10 plates in Explorer kit format.
  • Each reagent vial for Sample and Explorer kits is sufficient for one 96-, 384-, or 1536-well plate.
  • Suitable for endogenous, primary, and stem cell targets with improved signal window and assay robustness.